missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, BAKER INSTRA-ANALYZED Plus, J.T. Baker™

Product Code. 15588594 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
0.5 L
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15588594 0.5 L 500mL
15598594 1 L 1 L
15508604 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15588594 Supplier JT Baker Supplier No. 9385.0500
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Analyzed for up to 64 metals in the low parts-per-billion (ppb) range, ensuring low background interference for conducting critical trace metal analysis.

BAKER INSTRA-ANALYZED™ Plus acids allow for testing of more trace metals with tighter specifications.

  • Quality assured for up to 64 trace metals tested to low parts-per-billion (ppb) levels
  • Lot to lot consistency ensures reliable results between tests
  • Space-saving HDPE packaging with low metallic extractables
  • Enhanced safety with tamper-evident caps
  • Environmentally-friendly recyclable HDPE bottles
  • Strict protocols in an ISO facility ensure quality in manufacturing

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 0.1 (1N aq. soln.)
Color Colorless
Melting Point -35°C
Boiling Point 48°C
Density 1.18g/mL at 20°C
Odor Pungent
Packaging HDPE
Vapor Pressure 14.1 kPa
Assay Percent Range 34-37%
Quantity 0.5 L
Physical Form Liquid
Solubility Information Soluble in water
Formula Weight 36.46g/mol
Grade For use in Trace Metal Analysis
Assay w/w
Chemical Name or Material Hydrochloric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.