missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, AR™ (ACS), Macron Fine Chemicals™

Product Code. 15518774
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Polyethylene bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15518774 500 mL Polyethylene bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518774 Supplier Macron Fine Chemicals Supplier No. H61305

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An analytical-grade reagent of exceptional purity that is specially manufactured for standardizing volumetric solutions and preparing reference standards in laboratory applications.

Macron Fine Chemicals™ brand AR™ grade solvents achieve a combination of value and purity. These solvents meet ACS standards for analytical grade products, delivering reliable and consistent results.

  • Quality that ensures consistent results
  • Specifications that exceed AR grade requirements
  • Produced in ISO certified facilities to ensure reliability

Specifications

Appearance Passes test
pH <1 (at 20°C)
Color Colorless
CAS 7647-01-0
Melting Point 0°C
Boiling Point 50°C
Density 1.18kg/L
Heavy Metals (as Pb) Max. 0.1 ppm
Odor Pungent
Packaging Polyethylene bottle
Residue after Ignition Max. 4 ppm
Assay Percent Range 36.5 to 38.0%
Quantity 500 mL
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Fully miscible in water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Percent Purity 37%
Grade AR™ ACS
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.