missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 99.999999% (metals basis), 33% min

Product Code. 11315819
Click to view available options
Quantity:
25 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
25mL
This item is not returnable. View return policy

Product Code. 11315819

Brand: Thermo Scientific Alfa Aesar 010990.AU

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-01-0 | ClH | 36.46 g/mol

Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing. It finds use in the manufacture of polyvinyl chloride (PVC). Further, it is employed for regeneration of ion exchangers and in the tanning industry for the processing of leathers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Color Colorless to Yellow
Boiling Point 81.5°C to 110°C
Concentration ≤33%
Density 1.19 g/mL
Odor Pungent
Quantity 1000 mL
Physical Form Liquid, Double Distilled In Quartz
UN Number UN1789
Merck Index 14,4780
Solubility Information Miscible with water.
IUPAC Name hydrogen chloride
Formula Weight 36.46
Percent Purity 99.999999%
Assay (metals basis)
Chemical Name or Material Hydrochloric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.