Learn More
Hydrochloric Acid 6 M According to European Pharmacopoeia (EP), Reagecon
Fully factorized, high-purity, stable product developed and tested for titrations.
Brand: Reagecon 3001500-5L
Code : A0
Additional Details : Weight : 5.85000kg Transport : UN number : 1789 Chem class : 8 Pack group : II
Description
- European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified to a specification of ± 0.2%.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection as detailed in several EP monographs.
- NIST traceable.
Specifications
6 M | |
HDPE Bottle GL45 | |
5 L | |
Liquid |
Safety and Handling
- HYDROCHLORIC ACID 10% w/v
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid 37%