missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 6.0N Solution, Biotech Reagent, J.T. Baker™

Product Code. 15557814 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
19 L
Packaging:
Narrow Mouth Polyethylene Bottle
Poly Bottle
Unit Size:
1 L
19 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15557814 19 L Poly Bottle 19 L
14841332 1 L Narrow Mouth Polyethylene Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15557814 Supplier Avantor J.T.Baker Supplier No. 032707

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Packaging Poly Bottle
Specific Gravity 1L = 1.10kg
Quantity 19 L
Formula Weight 36.46
Grade Biotech Reagent
Chemical Name or Material Hydrochloric Acid, 6.0N Solution
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.