missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 5% v/v aq. soln.
Click to view available options
Quantity:
500 mL
1 L
4 L
Unit Size:
1 L
4 L
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Concentration | 5% v/v aq. soln. |
| Quantity | 4 L |
| Physical Form | Liquid |
| UN Number | UN1789 |
| IUPAC Name | palladium(2+) dichloride |
| Formula Weight | 36.46 |
| Chemical Name or Material | Hydrochloric acid |
Safety and Handling
Product Identifier
- Hydrochloric acid, 5% v/v aqueous solution
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : Hydrochloric acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction