missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 37%, extra pure, Thermo Scientific Chemicals

Product Code. 15242380
Click to view available options
Quantity:
5 L
Packaging:
Plastic Bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7647-01-0, 7732-18-5
Melting Point -35.0°C
Boiling Point 57.0°C
Density 1.1800g/mL
Packaging Plastic Bottle
Specific Gravity 1.18
Assay Percent Range 37%
Quantity 5 L
Molecular Formula ClH
Viscosity 1.9 mPa.s (15°C)
Linear Formula HCl
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46
Percent Purity 37%
Grade Extra Pure
Chemical Name or Material Hydrochloric Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid, 37%
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'