missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™

Catalog Number 10316380
61.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


35.15 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
7647-01-0
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
7647-01-0
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883

ClH, CAS Number-7647-01-0, spirits of salt, hydrogen chloride, hydrochloric acid, hydrogen chloride hcl, chlorohydric acid, chlorwasserstoff, chloorwaterstof, muriatic acid, acide chlorhydrique, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol

TRUSTED_SUSTAINABILITY
pH 1
Color Colorless
Melting Point -35°C
Boiling Point 57°C
Packaging HDPE plastic bottle
Vapor Pressure 125mbar at 20°C
Quantity 2.5 L
Viscosity 1.8 mPaS at 15°C
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
Formula Weight 36.46g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'