missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, meets analytical spec. of BP, NF, Ph. Eur., fuming, 36.5-38%, Solstice

Product Code. 15690840 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Paper / Poly Bag
Plastic Bottle
Unit Size:
1 L
2.5 L
30kg
Pack of 4
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15690840 6 x 1 L Plastic Bottle Pack of 6
15630840 1 L Glass Bottle 1 L
15650840 2.5 L Glass Bottle 2.5 L
15640840 2.5 L Plastic Bottle 2.5 L
15600850 6 x 1 L Glass Bottle Pack of 6
15680840 4 x 2.5 L Glass Bottle Pack of 4
15670840 4 x 2.5 L Plastic Bottle Pack of 4
15660840 30 kg Paper / Poly Bag 30kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15690840 Supplier Honeywell-Fluka Supplier No. 071026X1L
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Meets analytical specification of BP, NF, Ph. Eur., fuming, 36.5-38%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Color Yellow
Density 1.2g/mL at 25°C
Packaging Plastic Bottle
Residue after Ignition ≤0.002% (as SO4)
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 6 x 1 L
UN Number UN1789
IUPAC Name hydrogen chloride
Formula Weight 36.46
Percent Purity 36.5 to 38%
Chemical Name or Material Hydrochloric Acid, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.