missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid 34-37%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™

Product Code. 11355890 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355890 2.5 L 2.5 L
11365830 500 mL 500mL
11325870 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11355890 Supplier Fisher Chemical Supplier No. A508P212
  • ONLINE EXCLUSIVE PRICE
    £421.00
    £286.00 / 2.5 L
    Save £135.00
    32% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ClH, CAS Number-7647-01-0, 500mL, 57 deg.C, Colorless, 36.46g/mol, -35 deg.C, 11324, Liquid, 1.27, 125mbar at 20 deg.C, 1.8 mPaS at 15 deg.C | CAS: 7647-01-0 | ClH | 36.46 g/mol

ClH, CAS Number-7647-01-0, 500mL, 57 deg.C, Colorless, 36.46g/mol, -35 deg.C, 11324, Liquid, 1.27, 125mbar at 20 deg.C, 1.8 mPaS at 15 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 1
Color Colorless
Melting Point -35°C
Boiling Point 57°C
Vapor Pressure 125mbar at 20°C
Quantity 2.5 L
Viscosity 1.8 mPaS at 15°C
Physical Form Liquid
Formula Weight 36.46g/mol
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid, 34-37%
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.