missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid 30%, BAKER INSTRA-ANALYZED™, J.T. Baker™

Product Code. 13235503 Shop All J.T.Baker Products
Change view
Click to view available options
:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
13235503 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13235503 Supplier JT Baker Supplier No. 9600.1000PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Trace Metal Analysis

Specifications

Color Max. 10 (Hazen)
CAS 7647-01-0
Melting Point -30°C
Boiling Point 110°C (1013 hPa)
Density 1.18 g/cm³ (20°C)
Packaging Glass bottle
Assay Percent Range 29 to 31 %
Quantity 1 L
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
UN Number 1789
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1LT Hydrochloric Acid BAKER INSTRA-
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.