missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 2.0N Solution, Biotech Reagent, J.T. Baker™

Product Code. 15318521 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15318521 4 L Poly Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15318521 Supplier Avantor J.T.Baker Supplier No. 033603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Specifications

CAS 7647-01-0
Packaging Poly Bottle
Specific Gravity 1L = 1.03kg
Quantity 4 L
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Grade Biotech Reagent
Chemical Name or Material Hydrochloric Acid, 2.0N Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.