missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 1M, Eur. Grade, Reagecon™
BIOPHARMA

Product Code. 11474789 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
1L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11474789 1 L 1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474789 Supplier Reagecon Supplier No. 3001800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Concentration 1.0M
Packaging 1L
Quantity 1 L
compliance-icons
Product Identifier
  • HYDROCHLORIC ACID 1.0N (1.0M)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.