missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Management of Change (MOC) category = NI
- For Laboratory, Research, or Manufacturing Use
- Standardization at 25°C traceable to NIST Standard Reference Material
Specifications
Specifications
| Appearance | Passes Test |
| Appearance of Solution | Passes Test |
| CAS | 7647-01-0 |
| Heavy Metals | ≤ 0.5 ppm |
| Packaging | Plastic bottle |
| Normality | 0.995 - 1.005 |
| Quantity | 1 L |
| Molecular Formula | ClH |
| MDL Number | MFCD00011324 MFCD00792839 |
| UN Number | 1789 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?