missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
20 L
Description
Hydrochloric acid, 0.5N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Appearance | Colorless liquid |
Color | Colorless |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Quantity | 20 L |
Physical Form | Liquid |
Solubility Information | Miscible with water at room temperature. Also miscible with methanol,ethanol and ether. |
Formula Weight | 36.46 |
Concentration or Composition (by Analyte or Components) | Hydrochloric acid: 4.13%; Water: 95.87% |
Chemical Name or Material | Hydrochloric acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'