missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.5M, BAKER ANALYZED, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
Packaging:
Polyethylene (HDPE)
Unit Size:
1 L
Description
- Standardized potentiometrically against an NIST Standard Reference Material, Sodium Carbonate
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Specifications
Specifications
| Color | Colorless |
| CAS | 7647-01-0 |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Concentration | 0.4975 to 0.5025mol/L |
| Density | 1.0g/cm3 at 20°C |
| Heavy Metals (as Pb) | Max. 0.5 ppm |
| Packaging | Polyethylene (HDPE) |
| Vapor Pressure | 1.100 hPa (50°C) |
| Normality | 0.5N |
| Show More |
Safety and Handling
Signal Word
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction