missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.1M, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11484789 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Tamper evident
Förpackningsstorlek:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging unitSize
11484789 1 L Tamper evident 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11484789 Leverantör Reagecon Leverantörsnummer 3002100

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Manufactured according to Eur. Pharm. Chapter 4 (4.2.2) and prepared in an ISO 9001 facility. Hydrochloric Acid, 0.1M, EP Grade, Reagecon™ is a ready-to-use volumetric solution with 3.646g HCl per 1L water for use as a laboratory reagent.

Hydrochloric acid (HCl) is a corrosive, strong mineral acid with many uses, including as a laboratory reagent, in chloride production, as a neutralizing agent for bases, and as a catalyst and solvent in organic synthesis.

This solution is prepared by diluting 100mL of 1M hydrochloric acid in 1L water, where 1M hydrochloric acid is prepared by diluting 103g hydrochloric acid in 1L water.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.2.2) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise volumetric solutions

Specifikationer

pH 1
Color Colorless
CAS 7647-01-0, 7732-18-5
Boiling Point 100°C
Concentration 0.1M
Odor Odorless
Packaging Tamper evident
Vapor Pressure 190mmHg at 25°C
Certificate of Analysis Yes
Solvent or Matrix Water
Normality 0.1N
Quantity 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Soluble in water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Concentration or Composition (by Analyte or Components) 100mL of 1M hydrochloric acid diluted in 1L water
Chemical Name or Material Hydrogen chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.