missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 0.05N Standardized Solution

Product Code. 11353787
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Unit Size:
1 L
20 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353787 4 L 4 L
11343787 1 L 1 L
11333787 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353787 Supplier Thermo Scientific Alfa Aesar Supplier No. 035646.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7647-01-0 | ClH | 36.46 g/mol

Hydrochloric acid, 0.05N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
Solubility Miscible with water, methanol, ethanol and ether
IUPAC Name chlorane
Quantity 4 L
Formula Weight 36.46
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 0.413%; Water: 99.587%
Chemical Name or Material Hydrochloric acid, 0.05N

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.