missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.01M, Analytic Volumetric Solution, Reagecon™
Shop All Reagecon Products

Click to view available options
Quantity:
1 L
Packaging:
Bottle
Unit Size:
1 L
Description
Ready to use aqueous volumetric solution for all analytical laboratory applications, which can be used for manual or automatic titrations. This product is certified to a specification as outlined in the Certificate of Analysis. Lot Number and Expiry Date are printed on the label. Solution is certified traceable to N.I.S.T and this will be stated in the Certificate of Analysis.
- Molecular Formula: HCl
- Molar Mass: 36.46g/mol
- CAS: 7647-01-0
- EIN ECS No.: 231-595-7
Specifications
Specifications
| Color | Colorless |
| CAS | 7647-01-0 |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Concentration | 0.02mol/L |
| Density | 1.00049g/cm3 |
| Packaging | Bottle |
| Traceability to NIST | Traceable to NIST |
| Vapor Pressure | 23hPa (at 20°C) |
| Certificate of Analysis | Supplied |
| Show More |
Safety and Handling
Product Identifier
- HYDROCHLORIC ACID 0.01N (0.01M)
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction