missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocarbons Mixture (C6-C20) 595 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | n-Decane∣n-Dodecane∣n-Eicosane∣n-Hexadecane∣n-Hexane∣n-Octadecane∣n-Octane∣n-Tetradecane |
| CAS | 124-18-5, 112-40-3, 112-95-8, 544-76-3, 110-54-3, 593-45-3 |
| Analyte or Component Names | n-Decane∣n-Dodecane∣n-Eicosane∣n-Hexadecane∣n-Hexane∣n-Octadecane∣n-Octane∣n-Tetradecane |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | n-Decane 1000 μg/mL∣n-Dodecane 1000 μg/mL∣n-Eicosane 1000 μg/mL∣n-Hexadecane 1000 μg/mL∣n-Hexane 1000 μg/mL∣n-Octadecane 1000 μg/mL∣n-Octane 1000 μg/mL∣n-Tetradecane 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?