missing translation for 'onlineSavingsMsg'
Learn More

Hydrocarbons Mixture (C5-C32) 586 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30309787
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309787 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309787 Supplier Dr. Ehrenstorfer Supplier No. DREA50000586DI
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Heneicosane∣n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hentriacontane∣n-Heptacosane∣n-Heptadecane∣n-Heptane∣n-Hexacosane∣n-Hexadecane∣n-Hexane∣n-Nonacosane∣n-Nonadecane∣n-Nonane∣n-Octadecane∣n-Octane∣n-Pentacosane∣n-Pentadecane∣n-Pentane∣n-Tetradecane∣n-Tricosane∣n-Tridecane∣n-Undecane∣Octacosane∣Tetracosane∣Triacontane
CAS 629-94-7, 124-18-5, 629-97-0, 112-40-3, 544-85-4, 112-95-8
Analyte or Component Names Heneicosane∣n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hentriacontane∣n-Heptacosane∣n-Heptadecane∣n-Heptane∣n-Hexacosane∣n-Hexadecane∣n-Hexane∣n-Nonacosane∣n-Nonadecane∣n-Nonane∣n-Octadecane∣n-Octane∣n-Pentacosane∣n-Pentadecane∣n-Pentane∣n-Tetradecane∣n-Tricosane∣n-Tridecane∣n-Undecane∣Octacosane∣Tetracosane∣Triacontane
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Heneicosane 1000 μg/mL∣n-Decane 1000 μg/mL∣n-Docosane 1000 μg/mL∣n-Dodecane 1000 μg/mL∣n-Dotriacontane 1000 μg/mL∣n-Eicosane 1000 μg/mL∣n-Hentriacontane 1000 μg/mL∣n-Heptacosane 1000 μg/mL∣n-Heptadecane 1000 μg/mL∣n-Heptane 1000 μg/mL∣n-Hexacosane 1000 μg/mL∣n-Hexadecane 1000 μg/mL∣n-Hexane 1000 μg/mL∣n-Nonacosane 1000 μg/mL∣n-Nonadecane 1000 μg/mL∣n-Nonane 1000 μg/mL∣n-Octadecane 1000 μg/mL∣n-Octane 1000 μg/mL∣n-Pentacosane 1000 μg/mL∣n-Pentadecane 1000 μg/mL∣n-Pentane 1000 μg/mL∣n-Tetradecane 1000 μg/mL∣n-Tricosane 1000 μg/mL∣n-Tridecane 1000 μg/mL∣n-Undecane 1000 μg/mL∣Octacosane 1000 μg/mL∣Tetracosane 1000 μg/mL∣Triacontane 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.