missing translation for 'onlineSavingsMsg'
Learn More

Hydrocarbons Mixture (C10-C40) 545 1000 μg/mL in Heptane, Dr. Ehrenstorfer

Product Code. 30324984
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324984 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324984 Supplier Dr. Ehrenstorfer Supplier No. DREA50000545HP
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Octadecane∣n-Octatriacontane∣n-Tetradecane∣n-Tetratriacontane∣Octacosane∣Tetracontane∣Tetracosane∣Triacontane
CAS 124-18-5, 629-97-0, 112-40-3, 544-85-4, 112-95-8, 630-01-3
Analyte or Component Names n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Octadecane∣n-Octatriacontane∣n-Tetradecane∣n-Tetratriacontane∣Octacosane∣Tetracontane∣Tetracosane∣Triacontane
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Heptane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) n-Decane 1000 μg/mL∣n-Docosane 1000 μg/mL∣n-Dodecane 1000 μg/mL∣n-Dotriacontane 1000 μg/mL∣n-Eicosane 1000 μg/mL∣n-Hexacosane 1000 μg/mL∣n-Hexadecane 1000 μg/mL∣n-Hexatriacontane 1000 μg/mL∣n-Octadecane 1000 μg/mL∣n-Octatriacontane 1000 μg/mL∣n-Tetradecane 1000 μg/mL∣n-Tetratriacontane 1000 μg/mL∣Octacosane 1000 μg/mL∣Tetracontane 1000 μg/mL∣Tetracosane 1000 μg/mL∣Triacontane 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.