missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid Solution (in Acetic Acid), TRC

Product Code. 30394018
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Unit Size:
100mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394018 10 mL -
30395680 25 mL -
30391463 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394018 Supplier Toronto Research Chemicals Supplier No. TRCH71456510ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37348-16-6
Molecular Formula C2 H4 O2 . H Br
Molecular Weight (g/mol) 140.964
Synonym Acetic Acid Mixture with Hydrobromic Acid
IUPAC Name acetic acid;hydrobromide
SMILES Br.CC(=O)O

Specifications

Quantity 10 mL
InChI Formula InChI=1S/C2H4O2.BrH/c1-2(3)4;/h1H3,(H,3,4);1H
Formula Weight 139.9473
Chemical Name or Material Hydrobromic Acid Solution (in Acetic Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.