missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, purum p.a., Solstice

Product Code. 15661690 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15661690 250 mL 250mL
15651690 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15661690 Supplier Honeywell Research Chemicals Supplier No. 18730250ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purum p.a.,≥48%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name bromane
SMILES Br

Specifications

Density 1.49g/mL at 25°C
Vapor Pressure 8mmHg (25°C), 320psi (21.1°C)
Quantity 250 mL
UN Number 1788
Beilstein 3587158
Formula Weight 80.91g/mol
Percent Purity ≥48%
Grade purum p.a.
Chemical Name or Material Hydrobromic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.