missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, Purum p.a., ≥48%, Honeywell™

Product Code. 16665753 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
15 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16665753 15 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16665753 Supplier Honeywell Research Chemicals Supplier No. 1873015L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Light Yellow
CAS 7732-18-5
Melting Point -11°C
Boiling Point 126°C (1,013 hPa)
CAS Min % ≥ 50%
CAS Max % <70%
Density 1.490 g/cm3 (20°C)
Quantity 15 L
Molecular Formula H2O
Linear Formula HBr
Beilstein 3587158
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade Reagent
Chemical Name or Material Hydrobromic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.