Learn More
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Description
Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes, and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration, and dehydration, and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Appearance | Colorless to pale yellow liquid |
Color | Colorless to Yellow |
Density | 1.49 |
Sensitivity | Air Sensitive |
Vapor Pressure | 22500 hPa (16876mm Hg) at 20°C |
Assay Percent Range | 47.0-49.0% |
Quantity | 1 L |
Physical Form | Liquid |
Beilstein | 3587158 |
Solubility Information | Miscible in water. |
Show More |
RUO – Research Use Only