missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals

Catalog Number 11361588
Click to view available options
Quantity:
5 mL
100 mL
500 mL
This item is not returnable. View return policy
10035-10-6, 7732-18-5
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br
This item is not returnable. View return policy
10035-10-6, 7732-18-5
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br

Air Sensitive

Hydrobromic acid is used for the production of inorganic bromides of zinc, calcium, and sodium. It is used as a catalyst in some alkylation reactions. In the field of metallurgy, it is employed in the extraction of certain ores. Further, it is used as a reagent for the cleavage of certain ether linkages or as a brominating agent in organic transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Density 1.49 g/cm3
Sensitivity Air Sensitive
Vapor Pressure 22500 hPa (16876mm Hg)
Assay Percent Range 99.9999% (metals basis)
Quantity 100 mL
Physical Form Liquid
Beilstein 3587158
Solubility Information Miscible with water.
IUPAC Name hydrogen bromide
Formula Weight 80.92
Concentration or Composition (by Analyte or Components) Hydrogen bromide: 48.0%; Water: 52.0%
Chemical Name or Material Hydrobromic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'