missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln.

Product Code. 11325819
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325819 500 mL 500mL
11351588 5 mL 5mL
11361588 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11325819 Supplier Thermo Scientific Alfa Aesar Supplier No. 010991.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Air Sensitive | CAS: 10035-10-6 | BrH | 80.91 g/mol

Hydrobromic acid is used for the production of inorganic bromides of zinc, calcium, and sodium. It is used as a catalyst in some alkylation reactions. In the field of metallurgy, it is employed in the extraction of certain ores. Further, it is used as a reagent for the cleavage of certain ether linkages or as a brominating agent in organic transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Density 1.49
Sensitivity Air Sensitive
Vapor Pressure 22500 hPa (16876mm Hg)
Assay Percent Range 99.9999% (metals basis)
Quantity 500 mL
Physical Form Liquid
Beilstein 3587158
Solubility Information Miscible with water.
IUPAC Name bromane
Formula Weight 80.92
Concentration or Composition (by Analyte or Components) Hydrogen bromide: 48.0%; Water: 52.0%
Chemical Name or Material Hydrobromic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.