missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Hydrion pH Indicator Strips

Product Code. p-8004089 Shop All Cole-Parmer Products
Click to view available options
pH Range:
1 to 14
5 to 9
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11779899

Brand: Cole-Parmer™ 5920014

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cole-Parmer Hydrion pH Indicator Strips, pH Range: 1 to 14, Color Indicator: Yes, Format: Strip, For Use With: Immediate pH test, Includes: 100 Dip sticks and a handy matching color chart, Quantity: 100/Pk.

Specifications

Color Indicator Yes
pH Range 5 to 9
Format Strip
For Use With (Application) For immediate pH test
Includes 100 Dip sticks and a handy matching color chart
Quantity 100/Pk.
Type Hydrion pH Indicator Strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.