missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Hydrion pH Indicator Strips

Product Code. 11779899 Shop All Cole-Parmer Products
Change view
Click to view available options
pH Range:
1 to 14
5 to 9
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
11779899 5 to 9 Each
11765318 1 to 14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11779899 Supplier Cole-Parmer™ Supplier No. 5920014

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cole-Parmer Hydrion pH Indicator Strips, pH Range: 1 to 14, Color Indicator: Yes, Format: Strip, For Use With: Immediate pH test, Includes: 100 Dip sticks and a handy matching color chart, Quantity: 100/Pk.

Specifications

Color Indicator Yes
pH Range 5 to 9
Format Strip
For Use With (Application) For immediate pH test
Includes 100 Dip sticks and a handy matching color chart
Quantity 100/Pk.
Type Hydrion pH Indicator Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.