missing translation for 'onlineSavingsMsg'
Learn More

Hydrion Insta-Chek Mechanical pH Pencil

Product Code. 11785289 Shop All Cole-Parmer Products
Change view
Click to view available options
pH Range:
0 to 13
Unit Size:
Pack of 3
Product Code. pH Range unitSize
11785289 0 to 13 Pack of 3
1 options
This item is not returnable. View return policy

Product Code. 11785289

Brand: Hydrion 5920032

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Insta-Chek Mechanical pH Pencil immediately gives the surface reaction of any solid to the nearest pH value.

  • Retractable, nonrefillable, mechanical pencils are more convenient than paper-wrap pencils.
  • The widest range available—reading from pH 0 to 13—with 14 different color matches.
  • The handy, easily portable form is perfect for pH spot checks, requiring as little as 0.001 mL of buffered solution.
  • Use pencil on virtually any surface—even irregular or uneven ones.
  • It is effective on porous material and impervious solids.
  • To use, first wet the surface or area with distilled water and stroke several times with the pH pencil.
  • After 15 seconds, compare the resulting color against the color chart.
  • Pack includes three mechanical pencils and two color charts.

Specifications

pH Range 0 to 13
For Use With (Application) Immediately gives the surface reaction of any solid to the nearest pH value
Includes Three mechanical pencils and two color charts
Quantity 3 Pk.
Test Time 15 sec.
Type Hydrion Insta-Chek Mechanical pH Pencil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.