Learn More
Hydriodic acid, for analysis, 57 wt.% aqueous solution, distilled, unstabilized, Thermo Scientific Chemicals
£27.20 - £277.00
Chemical Identifiers
CAS | 10034-85-2 |
---|---|
Molecular Formula | HI |
Molecular Weight (g/mol) | 127.91 |
MDL Number | MFCD00011347 |
InChI Key | XMBWDFGMSWQBCA-UHFFFAOYSA-N |
IUPAC Name | hydrogen iodide |
SMILES | I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10002500
|
Thermo Scientific Acros
198360500 |
50 mL |
£27.20
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10441051
|
Thermo Scientific Acros
198362500 |
250 mL |
£75.60
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10013433
|
Thermo Scientific Acros
198360010 |
1 L |
£277.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10034-85-2 | |
127.91 | |
XMBWDFGMSWQBCA-UHFFFAOYSA-N | |
I |
HI | |
MFCD00011347 | |
hydrogen iodide |
Specifications
10034-85-2 | |
127.0°C | |
HI | |
MFCD00011347 | |
15, 4814 | |
Solubility aq. soln.: soluble. | |
I | |
127.91 | |
Analysis |
1.7000 g/mL | |
Glass bottle | |
HI | |
01,449; 02,213; 06,281; 07,173; 08,246; 09,238; 10,199; 11,255; 15,166 | |
1.7 | |
XMBWDFGMSWQBCA-UHFFFAOYSA-N | |
hydrogen iodide | |
127.91 | |
Hydriodic acid |
Safety and Handling
GHS H Statement Causes severe skin burns and eye damage.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-109-9
- Hydriodic acid, aqueous solution
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Hydriodic acid
RUO – Research Use Only