missing translation for 'onlineSavingsMsg'
Learn More

Hydrindantin dihydrate, 96%

Product Code. 10216210 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216210 50 g Glass bottle 50g
10521411 10 g Glass bottle 10g
10301344 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10216210 Supplier Thermo Scientific Acros Supplier No. 159280500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5950-69-6 | C18H14O8 | 358.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5950-69-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.29
MDL Number MFCD00149242
InChI Key QHVADKNWNMILPQ-HOTGVXAUSA-N
Synonym 2s-2,3,3-trihydroxy-2-2s-1,1,2-trihydroxy-3-oxoinden-2-yl inden-1-one
PubChem CID 6560392
IUPAC Name (2S)-2,3,3-trihydroxy-2-[(2S)-1,1,2-trihydroxy-3-oxoinden-2-yl]inden-1-one
SMILES C1=CC=C2C(=C1)C(=O)C(C2(O)O)(C3(C(=O)C4=CC=CC=C4C3(O)O)O)O

Specifications

Melting Point >240°C
Color Beige to Beige-Pink
Infrared Spectrum Authentic
Assay Percent Range 95% min
Packaging Glass bottle
Quantity 50 g
Merck Index 14, 4775
Solubility Information Solubility in water: insoluble
Formula Weight 358.29
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Hydrindantin dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Hydrindantin dihydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.