missing translation for 'onlineSavingsMsg'
Learn More

Hydridotetrakis(triphenylphosphine)rhodium(I)

Product Code. p-3212739 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10441723

Brand: Thermo Scientific Acros 377932500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18284-36-1 | C72H61P4Rh | 1153.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18284-36-1
Molecular Formula C72H61P4Rh
Molecular Weight (g/mol) 1153.08
MDL Number MFCD00015867
InChI Key QTTJMGBPWXLZKV-UHFFFAOYSA-N
Synonym hrh pph3 4, hydridotetrakis triphenylphosphine rhodium i, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium i hydride, molecular weight:456.19, molecular formula:c16h26o2rh2, cas number:73468-85-6, rhodium, tetrakis triphenylphosphine, hydridotetrakis triphenylphosphine rhodium
PubChem CID 11981874
IUPAC Name tetrakis(triphenylphosphane) rhodium
SMILES [RhH].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 140°C to 146°C
Color Ochre
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 mg
Formula Weight 1151.25
Physical Form Crystalline Powder
Chemical Name or Material Hydridotetrakis(triphenylphosphine)rhodium(I)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.