missing translation for 'onlineSavingsMsg'
Learn More

Hydridotetrakis(triphenylphosphine)rhodium(I)

Product Code. 10441723 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441723 250 mg Glass bottle 250mg
10695843 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441723 Supplier Thermo Scientific Acros Supplier No. 377932500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18284-36-1 | C72H61P4Rh | 1153.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18284-36-1
Molecular Formula C72H61P4Rh
Molecular Weight (g/mol) 1153.08
MDL Number MFCD00015867
InChI Key QTTJMGBPWXLZKV-UHFFFAOYSA-N
Synonym hrh pph3 4, hydridotetrakis triphenylphosphine rhodium i, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium i hydride, molecular weight:456.19, molecular formula:c16h26o2rh2, cas number:73468-85-6, rhodium, tetrakis triphenylphosphine, hydridotetrakis triphenylphosphine rhodium
PubChem CID 11981874
IUPAC Name tetrakis(triphenylphosphane) rhodium
SMILES [RhH].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 140°C to 146°C
Color Ochre
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 mg
Formula Weight 1151.25
Physical Form Crystalline Powder
Chemical Name or Material Hydridotetrakis(triphenylphosphine)rhodium(I)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.