missing translation for 'onlineSavingsMsg'
Learn More

Hydridotetrakis(triphenylphosphine)rhodium(I), Rh 8.9% min

Product Code. 11420650
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420650 5 g 5g
11410650 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420650 Supplier Thermo Scientific Alfa Aesar Supplier No. 044030.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18284-36-1 | C72H61P4Rh | 1153.08 g/mol

It finds its application as a catalyst for hydrogenation of alkenes and hydrosilylation of alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18284-36-1
Molecular Formula C72H61P4Rh
Molecular Weight (g/mol) 1153.08
MDL Number MFCD00015867
InChI Key QTTJMGBPWXLZKV-UHFFFAOYSA-N
Synonym hrh pph3 4, hydridotetrakis triphenylphosphine rhodium i, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium i hydride, molecular weight:456.19, molecular formula:c16h26o2rh2, cas number:73468-85-6, rhodium, tetrakis triphenylphosphine, hydridotetrakis triphenylphosphine rhodium
PubChem CID 11981874
SMILES [RhH].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 145°C to 147°C (decomposition)
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform, toluene
IUPAC Name tetrakis(triphenylphosphane) λ¹-rhodium
Formula Weight 1153.09
Concentration or Composition (by Analyte or Components) Rh 8.9% min
Physical Form Powder
Chemical Name or Material Hydridotetrakis (triphenylphosphine)rhodium (I)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.