missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine sulfate, 99+%

Product Code. 11439712
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439712 250 g 250g
11478880 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439712 Supplier Thermo Scientific Alfa Aesar Supplier No. A10792.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10034-93-2 | H6N2O4S | 130.118 g/mol

Hydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comIncompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-93-2
Molecular Formula H6N2O4S
Molecular Weight (g/mol) 130.118
MDL Number MFCD00044873
InChI Key ZGCHATBSUIJLRL-UHFFFAOYSA-N
Synonym hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate
PubChem CID 24842
IUPAC Name hydrazine;sulfuric acid
SMILES NN.OS(=O)(=O)O

Specifications

Melting Point 254°C
Density 1.37
pH 1.3
Odor Odorless
Quantity 250 g
UN Number UN3288
Merck Index 14,4772
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 130.12
Percent Purity ≥99%
Chemical Name or Material Hydrazine sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: hydrazine sulphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.