missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Hydrate, TRC

Product Code. 30007234
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007234 10 g 10g
30005955 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007234 Supplier Toronto Research Chemicals Supplier No. TRCH69760010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-57-8
Molecular Formula H2 O . H4 N2
Molecular Weight (g/mol) 50.06
Synonym Hydrazine, hydrate (1:1), Hydrazine, hydrate (6CI,7CI), Hydrazine, monohydrate (8CI,9CI), Hydrazine hydrate (1:1), Hydrazine hydrate, Hydrazine hydroxide, Hydrazinium hydroxide
IUPAC Name hydrazine;hydrate
SMILES O.NN

Specifications

Quantity 10 g
InChI Formula InChI=1S/H4N2.H2O/c1-2;/h1-2H2;1H2
Formula Weight 50.048
Percent Purity >95%
Chemical Name or Material Hydrazine Hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.