missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine hydrate, extra pure, solution 60%, SLR, Fisher Chemical

Product Code. 10141810 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
HDPE plastic bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10141810 500 mL HDPE plastic bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10141810 Supplier Fisher Chemical Supplier No. H/0500/PB08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7803-57-8
Melting Point -65°C
Color Colorless
pH 10.6 to 10.7
Boiling Point 109.4°C
Packaging HDPE plastic bottle
Molecular Formula H6N2O
MDL Number MFCD00149931
Quantity 500 mL
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydraziniumhydroxide, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
InChI Key IKDUDTNKRLTJSI-UHFFFAOYSA-N
SMILES O.NN
IUPAC Name hydrazine hydrate
Molecular Weight (g/mol) 50.06
PubChem CID 24654
Vapor Density 1.1 15
ChEBI CHEBI:35511
Formula Weight 32.04g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine hydrate solution 62%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Acute toxicity Category 2
  • Carcinogenicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Hydrazine hydrate, Hydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.