missing translation for 'onlineSavingsMsg'
Learn More

Hydrastine, MedChemExpress

Product Code. 16468638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468638 10 mg 10mg
16401088 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468638 Supplier Medchem Express Supplier No. HYB0927
 Restricted Item
Estimated Shipment: 09-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Hydrastine is a natural alkaloid which is present in Hydrastis canadensis and other plants of the ranunculaceae family.

Hydrastine is a natural alkaloid which is present in Hydrastis canadensis and other plants of the ranunculaceae family.

Specifikationer

Chemical Name or Material Hydrastine
CAS 118-08-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H21NO6
Quantity 10 mg
Synonym (-)-β-Hydrastine (1R,9S)-β-Hydrastine
Solubility Information DMSO : 100 mg/mL (260.83 mM; Need ultrasonic)
SMILES O=C1O[C@H]([C@@H]2N(C)CCC3=C2C=C(OCO4)C4=C3)C5=C1C(OC)=C(OC)C=C5
Molecular Weight (g/mol) 383.39
Formula Weight 383.39
Percent Purity 99.0%
Grade Research
Physical Form Solid
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.