missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Working Medium K, Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
Pack of 4
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™ - Working Medium K |
| Name Note | Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium) |
| Boiling Point | 60°C |
| CAS Min % | ≥50.0000%, ≥30.0000% |
| Density | ∼1.34 g/mL |
| CAS | 107-07-3 |
| Quantity | 4 x 1 L |
| CAS Max % | <50.0000% |
| Flash Point | 100°C |
| MDL Number | MFCD00147202 |
| Show More |
Safety and Handling
Product Identifier
- HYDRANAL-Working Medium K
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H300 + H310 + H330-Fatal if swallowed, in contact with skin or if inhaled.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : trichloromethane; chloroform, 2-chloroethanol; ethylene chlorohydrin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction