missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell™ Fluka™

Product Code. p-7122073 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
6 x (10 x 4mL), Ampules
Unit Size:
40mL
This item is not returnable. View return policy

Product Code. 15648380

Brand: Honeywell-Hydranal 3484740ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Density 0.86g/mL at 20°C
CAS 7732-18-5, 95-47-6
Quantity 6 x (10 x 4mL), Ampules
Flash Point 25°C (77°F)
UN Number UN1307
Chemical Name or Material HYDRANAL™-Water Standard 0.1
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : o-xylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.