missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 5, Honeywell Fluka™

Titrating agent for volumetric, two-component Karl Fischer titrations (Methanol-based)

Brand:  Honeywell Fluka 34801-6X1L-OM

Additional Details : CAS Number : 67-56-1 Weight : 5.10000kg Transport : UN number : 1230 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 16831910

  • £339.33 / 6 L
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Advantages of Hydranal™ Two-Component Reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents offer reduced toxicity compared to methanol
Specifications

Specifications

11°C
6 x 1 L
HYDRANAL™ - Titrant 5
Neutral
Metrohm OMNIS Smart Reagent
Liquid
Methyl Alcohol
Volumetric Two-Component KF Titration
Brown
0.850 g/cm3 (at 20°C)
7553-56-2
65°C (at 1.013 hPa)
-98°C
UN1230
Completely Miscible in Water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL NEXTGEN Titrant 5 OM

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, iodine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers