missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Titrant 2, Reagent for volumetric two-component Karl Fischer titration, Honeywell™ Fluka™
Shop All Honeywell Fluka ProductsSpecifications
Specifications
| Chemical Name or Material | HYDRANAL™-Titrant 2 |
| Density | 0.80g/mL |
| Quantity | 6 x 500 mL |
| Flash Point | 11°C (51.8°F) |
| UN Number | UN1230 |
Safety and Handling
Product Identifier
- HYDRANAL Titrant 2
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : methano; iodine
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?