missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 2 E, Reagent for volumetric two-component Karl Fischer titration (ethanol based), Honeywell™ Fluka™

Product Code. 15634360 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
Product Code. Quantity unitSize
15634360 6 x 1 L Pack of 6
15624360 1 L 1 L
2 options
This item is not returnable. View return policy

Product Code. 15634360

Brand: Honeywell-Hydranal 347236X1L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material HYDRANAL™ - Titrant 2 E
Name Note Reagent for volumetric two-component Karl Fischer titration (ethanol based)
Boiling Point 78°C
CAS Min % ≥1.0000%
CAS Max % <5.0000%, ≤100.0000%
CAS 68007-08-9, 7553-56-2
Density ∼0.83 g/mL
Quantity 6 x 1 L
Flash Point 12°C
Linear Formula H2O
MDL Number MFCD00011332
Packaging Glass Bottle
UN Number UN1170
Synonym Ethyl alcohol
Grade Karl Fischer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Titrant 2 E
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethanol; ethyl alcohol, iodine, 1H-Imidazole monohydriodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.