missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Standard sodium tartrate dihydrate, Standard for Karl Fischer titration (water content ∼15.66%), Honeywell™ Fluka™

Product Code. p-7122062 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
6 x 25 g
Unit Size:
25g
Pack of 6
This item is not returnable. View return policy

Product Code. 15644350

Brand: Honeywell-Hydranal 3469625G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Standard for Karl Fischer titration (water content ∼15.66%, exact value on report of analysis)

Advantages of HYDRANAL-Water Standards:

  • Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application
  • Manufactured according to ISO 9001 requirements
  • Report of Analysis showing the exact water content and expanded uncertainty

Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222

  • Long shelf life (up to five years)
  • Convenient packaging
  • Supplied with detailed instruction for use
TRUSTED_SUSTAINABILITY

Specifications

CAS 6106-24-7
Quantity 25 g
UN Number NONH for all modes of transport
Chemical Name or Material HYDRANAL™-Standard sodium tartrate dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.