missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Standard 5.0, Honeywell Fluka™

Product Code. 15664470 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL
6 x 500 mL
Unit Size:
100mL
500mL
Pack of 6
Product Code. Quantity unitSize
15664470 6 x 500 mL Pack of 6
15634470 100 mL 100mL
15644470 500 mL 500mL
15654470 6 x 100 mL Pack of 6
4 options
This item is not returnable. View return policy

Product Code. 15664470

Brand: Honeywell-Hydranal 348136X500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Test solution for rough check of volumetric KF equipment.

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Standard 5.0
CAS 71-36-3, 95-47-6
Quantity 6 x 500 mL
Flash Point 27°C (80.6°F)
UN Number UN1993C
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Standard 5.0
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : o-xylene, butan-1-ol; n-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.