missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Solver (Crude) oil, Solstice

Product Code. 15674350
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15674350 2.5 L 2.5 L
15664350 1 L 1 L
15694350 6 x 1 L Pack of 6
15684350 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15674350 Supplier Honeywell-Hydranal Supplier No. 346972.5L
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Medium for volumetric one- and two-component Karl Fischer titration in oils (chloroform-methanol-xylene-based)

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Solver (Crude) oil
CAS 67-56-1, 7446-09-5, 67-66-3, 95-47-6, 288-32-4
Density 1.01g/mL
Quantity 2.5 L
Flash Point 6°C (42.8°F)
UN Number UN1992
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Solver (Crude) oil
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: o-xylene, chloroform, methanol, imidazole, sulphur dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.