missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Sodium tartrate dihydrate, Honeywell Fluka™

Product Code. 15644430 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
6 x 100 g
Unit Size:
100g
25g
Pack of 6
This item is not returnable. View return policy

Product Code. 15644430

Brand: Honeywell-Hydranal 3480325G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Test substance for volumetric Karl Fischer titration (water content ∼15.66%, exact value on report of analysis)

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Sodium tartrate dihydrate
Quantity 25 g
UN Number NONH for all modes of transport
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.