missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - NEXTGEN Solvent E-FI, Honeywell Fluka™

Product Code. 16882360 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
6 L
This item is not returnable. View return policy

Product Code. 16882360

Brand: Honeywell Fluka 344311L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Medium for volumetric, two-component Karl Fischer titrations

  • Ethanol-based
  • Free of Imidazole
  • Free of CMR Substances
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - NEXTGEN Solvent E-FI
Melting Point -115°C
CAS Min % ≥55%
Color Colorless
Density 0.895 g/cm3
pH 5 to 6
Physical Form Liquid
CAS 64-17-5
Quantity 1 L
CAS Max % ≤80%
Packaging Glass Bottle
UN Number UN1170
Solubility Information Completely Miscible in Water
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.